Learn More
Keystone Safety Adjustable Cap Company Lab Coats
SDP
Catalog No. 19068428
Change view
Click to view available options
Size:
3X-Large
Catalog No. Size
19-068-428 3X-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-068-428 Supplier Keystone Safety Supplier No. LC0WENWHD3XLWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Product Type Controlled Environments Lab Coat
Size 3X-Large
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
No. of Pockets None
Latex Free Yes
Seam Type Serged