Learn More
Keystone Safety Isolation Gown
SDP
Catalog No. 19055315
This item is not returnable. View return policy
Catalog No. 19-055-315 Supplier Keystone Safety Supplier No. ISONWYELLOW
Add to Cart
Add to Cart
This item is not returnable. View return policy
Durable, multi-use, low-cost isolation gowns provide economical protection.
Non-woven, spunbond polypropylene material is breathable, lightweight and low-lint.
  • Protect against dirt, grime and non-hazardous dry particulates
  • Rear entry and extra-long ties
  • Mildew- and flame-resistant
  • Latex-free and non-allergenic
  • One size fits all up to 3X-Large
  • Color: Yellow
Ideal for food manufacturing, light industrial, medical or anywhere a low-particulate barrier garment is needed