Learn More
MSC Keystone Nylon Hairnet, Honeycomb, 24 in, Blue, Latex-Free, 100/Bag
Catalog No. 5000684596
Encompass
Keystone nylon honeycomb hairnets provide economical protection for controlled environments, such as Cleanroom Class 8. These hand-sewn, form-fitting hairnets offer comfortable and breathable wear, ensuring both user comfort and product integrity.
  • Made from 100% latex free nylon
  • Hand sewn for a comfortable, form-fitting design
  • Breathable and disposable
  • Strong and durable with non-linting properties
  • Suitable for cleanroom class 8 environments
  • FFDCA certified and Helmke drum tested
  • Features a 1/16 in hole size
Catalog No. 50-006-84596 Supplier MSC Supplier No. 73577033
Keystone nylon honeycomb hairnets provide economical protection for controlled environments, such as Cleanroom Class 8. These hand-sewn, form-fitting hairnets offer comfortable and breathable wear, ensuring both user comfort and product integrity.
  • Made from 100% latex free nylon
  • Hand sewn for a comfortable, form-fitting design
  • Breathable and disposable
  • Strong and durable with non-linting properties
  • Suitable for cleanroom class 8 environments
  • FFDCA certified and Helmke drum tested
  • Features a 1/16 in hole size