Learn More
Keystone Safety Polypropylene Arm Sleeves
SDP
Catalog No. 19068480
This item is not returnable. View return policy
Catalog No. 19-068-480 Supplier Keystone Safety Supplier No. AGNWIWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight, breathable fabric provides protection from dry particulates, dirt, and grime.
A high-quality, low-cost solution to protect workers against dry particulates, dirt and grime.
  • Made of non-woven, 100% spunbond polypropylene fabric
  • 100% latex free
  • White color
  • Flame resistant
  • 45.7 × 22.86cm (18 × 9 in.)
Specifications
Model AG-NWI-LARGE
Dimensions (L x W) 18 x 9 in. (45.7 x 22.8 cm)