Learn More
Specifications
Specifications
| pH | 7 |
| Melting Point | -95°C |
| Color | Colorless |
| Alkalinity | 10mg/kg max. (FCC) |
| Decomposition Information | >4°C |
| Density | 0.790g/cm³ |
| Quantity | 200 L |
| Identification | Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF) |
| Packaging | Steel Drum |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. NFPA Health:1 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.