Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, Certified ACS Plus, 36.5 to 38.0%, Fisher Chemical™
GSA_VA

Catalog No. A144C212 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
19 L
6.5 gal.
6 x 2.5 L
500 lb.
Packaging:
Glass Bottle
Glass Carboy
N/A
Poly Drum
Safe-Cote™ Glass
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A144C-212 6 x 2.5 L Glass Bottle
A144-500 500 mL Glass Bottle
A144S-500 500 mL Safe-Cote™ Glass
A144-212 2.5 L Glass Bottle
A144SI-212 2.5 L Safe-Cote™ Glass
A144S-212 6 x 2.5 L Safe-Cote™ Glass
A144P-19 19 L N/A
A144-500LB 500 lb. Poly Drum
A144-612GAL 6.5 gal. Glass Carboy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. A144C-212
Add to Cart

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH <1
Color Colorless
Melting Point -35°C
Boiling Point 57°C
Density 1.18g/cm³
Heavy Metals (as Pb) 0.1ppm max.
Identification Pass Test
Packaging Glass Bottle
Residue after Ignition 4ppm max.
Specific Gravity 1.18
Vapor Pressure 125mbar at 20°C
Quantity 6 x 2.5 L
Physical Form Liquid
Solubility Information Soluble in water
Percent Purity 36.5 to 38.0% (w/w)
Show More Show Less
Safety handling Solvent
missing translation for 'healthHazard2' DANGER!
missing translation for 'healthHazard3' Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:3
Flammability:0
Instability:1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.