Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone (Certified ACS), Fisher Chemical™
GSA_VA

Catalog No. A18200 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Aluminum Can
Glass Bottle
HDPE Bottle
Poly Bottle
Steel Drum
Steel Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A18-200 200 L Steel Drum
A18-1 1 L HDPE Bottle
A18-4 4 L Glass Bottle
A18P-4 4 L Poly Bottle
A18-20 20 L Steel Pail
A18-500 500 mL HDPE Bottle
A18S-4 4 L Aluminum Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. A18-200
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH 7
Melting Point -95°C
Color Colorless
Decomposition Information >4°C
Density 0.790g/cm³
Quantity 200 L
Flash Point -20°C
Identification Pass Test
Infrared Spectrum Conforms to reference
Packaging Steel Drum
Physical Form Liquid
Residue after Evaporation 0.001% max.
Specific Gravity 0.79
Assay Percent Range Assay: ≥99.5%
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0006mEq/g max.
Vapor Pressure 247mbar at 20°C
Viscosity 0.32 mPa/s at 20°C
UN Number 1090
Water 0.5% max.
Merck Index 15, 65
Boiling Point 56°C
Formula Weight 58.08
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Acetone
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.