Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial (Certified ACS), Fisher Chemical™

Catalog No. A38450LB Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
2.5 L
20 L
6 x 2.5 L
450 lb.
Packaging:
Glass Bottle
Poly Drum
Safe-Cote™ Glass

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Assay Percent Range Assay: ≥99.7% w/w
Linear Formula CH3COOH
UN Number 2789
Merck Index 15, 54
Solubility Information Soluble in water
Formula Weight 60.05
Percent Purity ≥99.7% (w/w)
Grade Certified ACS
Density 1.048g/cm³
Quantity 450 lb.
Flash Point 40°C
Packaging Poly Drum
Physical Form Liquid
Residue after Evaporation 8ppm max.
Vapor Pressure 1.52 kPa at 20°C
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:2
Instability:0
DOTInformation DOT Class 8: Corrosive
Recommended Storage RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.