Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid, Glacial (Certified ACS), Fisher Chemical
Shop All Fisherbrand Products

Click to view available options
:
Glass Bottle/Safe-Cote Individual; 2.5L
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| pH | 2.5 |
| Melting Point | 16°C |
| CAS | 64-19-7 |
| Boiling Point | 117°C |
| Color | Colorless |
| Assay Percent Range | Assay: ≥99.7% w/w |
| Molecular Formula | C2H4O2 |
| Linear Formula | CH3COOH |
| MDL Number | MFCD00036152 |
| UN Number | 2789 |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting. NFPA Health:3 Flammability:2 Instability:0 |
| DOTInformation | DOT Class 8: Corrosive |
| Recommended Storage | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction