Learn More
Ethyl Alcohol Denatured (Proprietary Solvent), Certified, Fisher Chemical™
Shop All Fisherbrand Products

Description
Formulation: Ethyl Alcohol (100 parts), methyl alcohol (at least 4 parts), methyl iso-butyl ketone (at least 1 part), ethyl acetate (at least 0.87 parts), and rubber hydrocarbon solvent (at least 1 part)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Melting Point | -90.0°C |
| Color | Colorless |
| Density | 0.7905g/cm³ |
| Quantity | 20 L |
| Flash Point | 13.9°C (57°F) |
| Identification | Pass Test |
| Packaging | Lined Steel Drum |
| Physical Form | Liquid |
| Specific Gravity | 0.7905 |
| Vapor Pressure | 40.9mmHg at 20°C |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Harmful by inhalation, in contact with skin and if swallowed. Exposure through inhalation may result in delayed pulmonary edema, which may be fatal. Aspiration hazard if swallowed - can enter lungs and cause damage. Possible risks of irreversible effects. Causes respiratory tract burns. Irritating to eyes and skin. May cause central nervous system effects. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Oxidizing agent. NFPA Health:2 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.