Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
GSA_VA

Catalog No. A461212 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A461-212 2.5 L Glass Bottle
A461-500 500 mL Glass Bottle
A461-1 1 L Wide Neck Amber Bottle
A461-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A461-212
  • $434.00
    $287.65 / Each

    Save $146.35

    34% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,333.00
    $883.65 / Case of 4 EA

    Save $449.35

    34% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, isopropanol, 2-hydroxypropane, isopropyl alcohol, alkolave, hartosol, dimethylcarbinol, avantine, sec-propyl alcohol, petrohol, 2-propanol, 2.5L, Report AU at 254nm, Report AU at 230nm, Report AU at 210nm, Report AU at 220nm | CAS: 67-63-0 | C3H8O | 60.096 g/mol

2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number 11674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH 7
Melting Point -89.5°C
Absorbance Report AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm
Color Colorless
Density 0.785g/cm³
Quantity 2.5 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 1ppm max.
Assay Percent Range Assay: ≥99.90%
Vapor Pressure 43mmHg at 20°C
Linear Formula (CH3)2CHOH
Viscosity 2.27 mPaS at 20°C
Water 0.05% max.
Merck Index 15, 5254
Boiling Point 81°C
Formula Weight 60.1
Percent Purity ≥99.90%
Grade Also meets ACS specifications
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.