Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
GSA_VA

Catalog No. A4671 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2 L
Packaging:
FEP Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A467-1 1 L FEP Bottle
A467-250 250 mL FEP Bottle
A467-500 500 mL FEP Bottle
A467-2 2 L FEP Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A467-1
Only null left
Add to Cart

Nitric Acid, HNO3, CAS Number-7697-37-2, acide nitrique, acidum nitricum, azotic acid, rfna, aqua fortis, nitryl hydroxide, salpetersaeure, nitrous fumes, nital, hydrogen nitrate, 500mL

  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially manufactured and cleaned FEP fluoropolymer bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH 1
Color Colorless Yellow
Melting Point -41°C
Boiling Point 120.5°C
Density 1.40 g/cm3
Packaging FEP Bottle
Vapor Pressure 0.94kPa (20°C)
Certificate of Analysis Certificate of Lot Analysis is included with each bottle.
Quantity 1 L
Physical Form Liquid
UN Number 2031
Merck Index 15, 6663
Formula Weight 63.01
Grade OPTIMA
Chemical Name or Material Nitric Acid
Show More Show Less
DOTInformation DOT Class 8: Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.