Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Propanol (USP), Fisher Chemical™
Shop All Fisherbrand Products

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Glass Bottle
Steel Drum
Steel Pail
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| pH | 7 |
| Melting Point | -89.5°C |
| Color | Colorless |
| Acidity | Pass Test |
| Density | 0.785g/cm³ |
| Boiling Point | 81°C |
| Quantity | 4 L |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen. Note: Products are tested to meet the analytical requirements of the noted grade (USP, Ph. EUR or otherwise). Product test results are available on the Certificate of Analysis or upon request for the specific product. Products are for research use or further manufacturing. Products are not for direct administration to humans or animals. It is the responsibility of the final formulator and end-user to qualify, test and/or validate each product for its intended use as an ingredient in the end product and in accordance with applicable regulations (including without limitation WHO-recommended Hand Rub Formulations (Hand Sanitizers) and/or FDA Policy for Preparation of Certain Alcohol-based Hand Sanitizer Products, as updated). Thermo Fisher Scientific expressly disclaims all non-specified warranties, expressed or implied, including the implied warranties of merchantability and fitness for a particular use. NFPA Health:1 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction