Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone (NF/FCC/EP), Fisher Chemical™

Catalog No. A9200 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
200 L
Packaging:
EcoSafPak™ Glass Bottle
Steel Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A9-200 200 L Steel Drum
A9-4 4 L EcoSafPak™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A9-200

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH 7
Melting Point -95°C
Color Colorless
Alkalinity 10mg/kg max. (FCC)
Appearance Pass Test (EP)
Decomposition Information >4°C
Density 0.790g/cm³
Quantity 200 L
Identification Pass Test, Pass Test (EP), Pass Test (FCC), Pass Test (NF)
Packaging Steel Drum
Physical Form Liquid
Refractive Index 1.358 to 1.360 (at 25°C), 1.358 to 1.360 (at 25°C) (FCC)
Residue after Evaporation 50ppm max. (EP)
Specific Gravity 0.789 Max. (NF), 0.7880 max. (FCC)
Vapor Pressure 247mbar at 20°C
Linear Formula CH3COCH3
Viscosity 0.32 mPa/s at 20°C
Water ≤3g/L (EP), 0.5% max. (FCC), 0.5% max. (NF)
UN Number 1090; Colorless
Boiling Point 56°C
Formula Weight 58.08
Percent Purity ≥99.0% (NF)
Grade BP/EP/FCC/JP/NF/USP
Chemical Name or Material Acetone
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.