Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Optima™, for HPLC and GC, Fisher Chemical™
GSA_VA

Catalog No. A9294 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Soda Lime Glass
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A929-4 4 L Amber Soda Lime Glass
A929-1 1 L Wide Neck Amber Bottle
A929SK-4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A929-4
  • $479.50
    $256.65 / Each

    Save $222.85

    46% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,436.00
    $768.65 / Case of 4 EA

    Save $667.35

    46% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 300-400nm range, to reduce interferences for smoother baselines
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH 7
Melting Point -95°C
Absorbance 1.00 AU max. 330nm, 0.06 AU max. at 340nm, 0.010 AU max. 350nm, 0.01 AU max. (350 to 400nm)
Color Colorless
Decomposition Information >4°C
Density 0.790g/cm³
Quantity 4 L
Flash Point -20°C (-4°C)
Identification Pass Test
Packaging Amber Soda Lime Glass
Physical Form Liquid
Refractive Index 1.3555 to 1.3580 (at 25°C)
Residue after Evaporation 1ppm max.
Assay Percent Range 99.9% min by GC
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0006mEq/g max.
Vapor Pressure 247mbar at 20°C
Linear Formula CH3COCH3
Viscosity 0.32 mPa/s at 20°C
Water 0.5% max.
UN Number 1090
Boiling Point 56°C
Formula Weight 58.08
Percent Purity ≥99.5%
Grade High purity mobile phase for HPLC and/or extraction solvent for GC applications
Chemical Name or Material Acetone
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.