Learn More
Isobutylamine
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -85.0°C |
| Color | Colorless to Yellow |
| Density | 0.7300g/mL |
| Boiling Point | 64.0°C to 71.0°C |
| Flash Point | −9°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Refractive Index | 1.397 |
| Linear Formula | (CH3)2CHCH2NH2 |
| Quantity | 2.5 L |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for |
| EINECSNumber | 201-145-4 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.