Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide, 0.1 N standard solution

Catalog No. AC124190025
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Plastic Bottle
Plastic Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC124190025 2.5 L Plastic Bottle
AC124190010 1 L Plastic Bottle
AC124190250 25 L Plastic Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC124190025 Supplier Thermo Scientific Chemicals Supplier No. 124190025
Only null left
Add to Cart

Sodium hydroxide, HNaO, CAS Number-1310-73-2, 7732-18-5, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, CHEBI:32145, Beige to White, 0.0995 to 0.1005N (20 deg.C), 1.0000g/mL | CAS: 1310-73-2 | HNaO | 39.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium hydroxide
Color White to Beige
Name Note 0.1 N Standard Solution
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Linear Formula NaOH
Concentration 0.0995 to 0.1005N (20°C)
MDL Number MFCD00003548
Fieser 01,1083; 05,616; 07,336; 08,460
Merck Index 15, 8761
Density 1.0000g/mL
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol
Quantity 2.5 L
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40
Grade Pure
Packaging Plastic Bottle
Specific Gravity 1
Physical Form Crystalline Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.