Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid, 1N standard solution
GSA_VA

Catalog No. AC124210025
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Plastic Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC124210025 2.5 L Plastic Bottle
AC124210010 1 L Plastic Bottle
AC124210100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC124210025 Supplier Thermo Scientific Chemicals Supplier No. 124210025
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrochloric acid
Color Colorless
Name Note 1N standard solution
CAS 7647-01-0, 7732-18-5
Concentration 0.998 to 1.002N (20°C)
Density 1.0200g/mL
Packaging Plastic Bottle
Quantity 2.5 L
Molecular Formula ClH
Physical Form Liquid
Linear Formula HCl
MDL Number MFCD00011324 MFCD00792839
Fieser 04,450; 05,533; 06,283
Merck Index 15, 4818
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.45
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.