Learn More
Hydrochloric acid, for analysis, fuming, 37% solution in water
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -35.0°C |
| Boiling Point | 57.0°C |
| Density | 1.1800g/mL |
| Heavy Metals (as Pb) | 2ppm max. |
| Packaging | Glass Bottle |
| Reducing Substances | (SO2) Passes Test |
| Specific Gravity | 1.18 |
| Quantity | 1 L |
| Viscosity | 1.9 mPa.s (15°C) |
| Physical Form | Solution |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. May be corrosive to metals. |
| Precautionary Statement | GHS P Statement Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep only in original container. Absorb spillage to prevent material damage. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.