Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dichlorobutyrophenone, 97%, Thermo Scientific™

Catalog No. AC136891000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC136891000 100 g
AC136890250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC136891000 Supplier Thermo Scientific Chemicals Supplier No. 136891000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40877-09-6
Molecular Formula C10H10Cl2O
Molecular Weight (g/mol) 217.09
MDL Number MFCD00018993
InChI Key UKCHLVFIVJBCKE-UHFFFAOYSA-N
Synonym 4-chloro-1-4-chlorophenyl butan-1-one, 4,4'-dichlorobutyrophenone, 1-butanone, 4-chloro-1-4-chlorophenyl, gamma,p-dichlorobutyrophenone, 4-chloro-1-4-chlorophenyl-1-oxobutane, 4-chloro-1-4-chlorophenyl-1-butanone, acmc-20a5su, 4',gamma-dichlorobutyrophenone, 4-chloro-4'-chlorobutyrophenone, 4-chloro-4'-chloro-butyrophenone
PubChem CID 2723781
IUPAC Name 4-chloro-1-(4-chlorophenyl)butan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCCl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 29°C to 30°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula ClC6H4CO(CH2)3Cl
Quantity 100 g
Solubility Information Solubility in water: insoluble
Formula Weight 217.09
Percent Purity 97%
Physical Form Pellets
Chemical Name or Material 4, 4'-Dichlorobutyrophenone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 255-123-4
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.