Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-1,2-propanediol, 98%

Catalog No. AC148120010
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148120010 1 L Glass bottle
AC148120250 25 mL Glass bottle
AC148125000 500 mL Glass bottle
AC148120025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC148120010 Supplier Thermo Scientific Chemicals Supplier No. 148120010
Only null left
Add to Cart

CAS: 96-24-2 | C3H7ClO2 | 110.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-24-2
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.54
MDL Number MFCD00004712
InChI Key SSZWWUDQMAHNAQ-UHFFFAOYNA-N
Synonym 3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin
PubChem CID 7290
ChEBI CHEBI:18721
IUPAC Name 3-chloropropane-1,2-diol
SMILES OCC(O)CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -40.0°C
Color Yellow
Density 1.3220g/mL
Boiling Point 213.0°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4790 to 1.4810
Linear Formula ClCH2CH(OH)CH2OH
Quantity 1 L
Specific Gravity 1.322
Merck Index 15, 2146
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,methanol and ether,slightly soluble in toluene
Viscosity 215 mPa.s (20°C)
Formula Weight 110.54
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Chloro-1, 2-propanediol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing genetic defects.
Fatal if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Fatal if inhaled.
May cause respiratory irritation.
Causes skin irritation.<br
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
EINECSNumber 202-492-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.