Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane, 99+%, for analysis

Catalog No. AC160780025
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160780025 2.5 L Glass Bottle
AC160780010 1 L Glass Bottle
AC160780251 25 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC160780025 Supplier Thermo Scientific Chemicals Supplier No. 160780025
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 110-54-3 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95.0°C
Color Colorless to Yellow
CAS Min % 99.0
CAS Max % 100.0
Density 0.6590g/mL
Quantity 2.5 L
Flash Point −22°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3748 to 1.3810
Assay Percent Range 99+%
Residue after Evaporation 0.002 % max.
Specific Gravity 0.659
Linear Formula CH3(CH2)4CH3
Vapor Pressure Vapor Pressure: 160mbar at 20°C
UN Number 1208
Viscosity 0.31 mPa.s (20°C)
Beilstein 01, 142
Water 0.02% Max. (Coulometric)
Merck Index 15, 4732
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Boiling Point 69.0°C
Formula Weight 86.18
Percent Purity ≥99%
Grade Analytical
Chemical Name or Material n-Hexane
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.