Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, 99+%, for spectroscopy

Catalog No. AC167900025
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167900025 2.5 L Glass Bottle
AC167900010 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167900025 Supplier Thermo Scientific Chemicals Supplier No. 167900025
  • $208.00
    $162.65 / Each of 1

    Save $45.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
 Restricted Item
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 108-88-3 | C7H8 | 92.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Absorbance (1 cm cell vs water), 0.01 max. at 350nm, 0.02 max. at 335nm, 0.07 max. at 310nm, 0.1 max. at 300nm, 0.4 max. at 288nm, 1 max. at 285nm
Density 0.8660g/mL
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range 99+%
Vapor Pressure Vapor Pressure: 29mbar at 20°C
Linear Formula C6H5CH3
Water 0.03% max.
Beilstein 05, 280
Merck Index 15, 9688
Formula Weight 92.14
Percent Purity ≥99.0%
Grade Spectroscopy
Chemical Name or Material Toluene, 99+%
Show More Show Less
Hazard Category Signal Word: DANGER!
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
EINECSNumber 203-625-9
RTECSNumber XS5250000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.