Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol, 99.5%, for analysis

Catalog No. AC184130010
Change view
Click to view available options
Quantity:
1 L
10 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Metal Drum
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC184130010 1 L EcoSafPak™ Glass Bottle
AC184130100 10 L Plastic Drum
AC184130250 25 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC184130010 Supplier Thermo Scientific Chemicals Supplier No. 184130010
  • $120.60
    $102.65 / Each

    Save $17.95

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $371.00
    $315.65 / Case of 4 EA

    Save $55.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67-63-0 | C3H8O | 60.096 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -89.5°C
Absorbance 0.01 max. (EurPh) at 310nm, 0.02 max. (EurPh) at 290nm, 0.03 max. (EurPh) at 270nm, 0.1 max. (EurPh) at 250nm, 0.3 max. (EurPh) at 230nm
Color Colorless
Density 0.7850g/mL
Quantity 1 L
Flash Point 12°C
Infrared Spectrum Authentic
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3760 to 1.3780
Specific Gravity 0.785
Assay Percent Range 99.5%
Vapor Pressure Vapor Pressure: 43mmHg at 20°C
Linear Formula (CH3)2CHOH
Viscosity 2.27 mPa.s (20°C)
Water 0.5% max.
Beilstein 01, 360
Fieser 13,108
Merck Index 15, 5254
Boiling Point 81.0°C to 83.0°C
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride,, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
Formula Weight 60.1
Percent Purity 99.5%
Grade Analytical
Chemical Name or Material Isopropanol, Reag. Ph. Eur.
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.