Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(propylene glycol), average M.W. 4.000

Catalog No. AC192202500
Click to view available options
Quantity:
250 g
500 g
Packaging:
Glass bottle

Catalog No. AC192202500

Supplier: Thermo Scientific Chemicals 192202500

Only null left
Add to Cart

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO

Specifications

Color Green to Yellow
Physical Form Viscous Liquid
Linear Formula H[OCH(CH3)CH2]nOH
Density 1.0000g/mL
Quantity 250 g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4500 to 1.4520
Specific Gravity 1
Viscosity 930 mm2/s (25°C)
Chemical Name or Material Poly(propylene glycol), Average M.W. 4.000
Show More Show Less
RTECSNumber OE5784360
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.