Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                100 mL
                            
                        
                            
                                500 mL
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
        
             
            Specifications
Specifications
| Chemical Name or Material | Cobalt standard solution | 
| Name Note | 1 mg/ml Co in 2% HNO3 | 
| Packaging | Plastic bottle | 
| CAS | 10141-05-6, 7732-18-5 | 
| Physical Form | Solution | 
| Molecular Formula | CoN2O6 | 
| Concentration | 990 to 1010 μg/mL of Co(2+) in 2 to 5% HNO3 | 
| MDL Number | MFCD00010935 | 
| Solubility | Miscible with water | 
| InChI Key | UFMZWBIQTDUYBN-UHFFFAOYSA-N | 
| Show More | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                