Learn More
Potassium dichromate, 99.5%, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 500.0°C |
| Melting Point | 398.0°C |
| Color | Orange to Orange-Red |
| Physical Form | Crystals |
| Quantity | 1 kg |
| Assay Percent Range | 99.4% min. (Iodometry) |
| Linear Formula | K2Cr2O7 |
| Fieser | 08,410; 09,383; 10,324; 12,405 |
| Merck Index | 15, 7748 |
| Solubility Information | Solubility in water: 125g/L (20°C). Other solubilities: 50 g/100 mL water (100°C), insoluble in alcohol |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. Fatal if inhaled. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. May cause allergy or asthma symptoms or breathing difficu |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. In case of major |
| EINECSNumber | 231-906-6 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.