Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium orthovanadate, 99%

Catalog No. AC205332500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205332500 250 g Plastic bottle
AC205330500 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC205332500 Supplier Thermo Scientific Chemicals Supplier No. 205332500
Only null left
Add to Cart

CAS: 13721-39-6 | Na3O4V | 183.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13721-39-6
Molecular Formula Na3O4V
Molecular Weight (g/mol) 183.91
MDL Number MFCD00003511
InChI Key IHIXIJGXTJIKRB-UHFFFAOYSA-N
Synonym sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
PubChem CID 61671
ChEBI CHEBI:35607
IUPAC Name trisodium;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Melting Point 850.0°C to 866.0°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 98.5% min. (as Na3VO4)
Linear Formula Na3VO4
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in ethanol
Formula Weight 183.91
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Sodium orthovanadate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 237-287-9
RTECSNumber YW1120000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.