Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver hexafluorophosphate, 98%, -6 mesh

Catalog No. AC211120250
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Plastic bottle

Catalog No. AC211120250

Supplier: Thermo Scientific Chemicals 211120250

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

CAS: 26042-63-7 | AgF6P | 252.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26042-63-7
Molecular Formula AgF6P
Molecular Weight (g/mol) 252.83
MDL Number MFCD00003415
InChI Key SCQBROMTFBBDHF-UHFFFAOYSA-N
Synonym silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
PubChem CID 168464
IUPAC Name silver;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[Ag+]

Specifications

Color Beige-Gray to White
Physical Form Powder
Quantity 25 g
Linear Formula AgPF6
Solubility Information Solubility in water: soluble
Formula Weight 252.83
Percent Purity 98%
Packaging Plastic bottle
Chemical Name or Material Silver hexafluorophosphate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 247-428-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.