Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mercury(I) nitrate dihydrate, 98%, for analysis
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 70.0°C |
| Color | White to Yellow |
| Physical Form | Crystalline Powder |
| Quantity | 50 g |
| Assay Percent Range | 97.5% min |
| Linear Formula | Hg2(NO3)2·2H2O |
| Fieser | 06,360 |
| Merck Index | 13, 5923 |
| Solubility Information | Solubility in water: 20g/L (20°C). |
| Formula Weight | 561.22 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Very toxic to aquatic life with long lasting effects. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash w |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction