Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum chloride hexahydrate, 99%, pure
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid

Specifications
Specifications
Quantity | 1 kg |
Assay Percent Range | 98.5% min. (Argentometry) |
Linear Formula | AlCl3·6H2O |
Fieser | 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 08,13; 09,11; 17,15 |
Merck Index | 15,333 |
Solubility Information | Solubility in water: 477g/L (20°C). Other solubilities: soluble in alcohol, ether, glycerol and propylene |
IUPAC Name | aluminium(3+) hexahydrate trichloride |
Formula Weight | 241.43 |
Percent Purity | 99% |
Grade | Pure |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. If eye irritation persists: Get medical advice/attention. If skin irritation occurs: Get medical advice/attention. Wash face,hands and any ex |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction