Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid, 99.8%, for analysis
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
20 L
Packaging:
Glass Bottle
Plastic Can
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Specifications
| Melting Point | 16.0°C to 16.5°C |
| Boiling Point | 117.0°C to 118.0°C |
| CAS Min % | 99.75 |
| CAS Max % | 100.0 |
| Assay Percent Range | 99.8% |
| Linear Formula | CH3COOH |
| UN Number | 2789 |
| Beilstein | 02, 96 |
| Fieser | 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12 |
| Merck Index | 15, 54 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS P Statement Causes severe skin burns and eye damage. Flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction