Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-hexane, 95+%, for HPLC
GREENER_CHOICE

Catalog No. AC232100010
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232100010 1 L Glass Bottle
AC232100025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC232100010 Supplier Thermo Scientific Chemicals Supplier No. 232100010
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 110-54-3, C6H14, 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95.0°C
Absorbance 1 cm cell vs HPLC-grade water, 1 cm cell vs HPLC-grade water at 0 nm, 0.005 max. at 250nm, 0.01 max. at 240nm, 0.03 max. at 230nm, 0.07 max. at 220nm, 0.25 max. at 210nm, 0.7 max. at 200nm
CAS Min % 95.0
Color Colorless
CAS Max % 100.0
Acidity 0.0002 meq/g max.
Density 0.6590g/mL
Quantity 1 L
Flash Point -22°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3748 to 1.3810
Residue after Evaporation 0.0005% max.
Assay Percent Range 95+%
Specific Gravity 0.659
Linear Formula CH3(CH2)4CH3
Viscosity 0.31 mPa.s (20°C)
UN Number 1208
Water 0.01% max.
Green Features ACT Ecolabel (Silver)
Boiling Point 69.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Formula Weight 86.18
Percent Purity ≥95%
Grade HPLC
Chemical Name or Material n-Hexane, For HPLC
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.