Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptopyridine-N-oxide, sodium salt, 40 w/w % aqueous solution

Catalog No. AC257740025
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC257740025 2.5 L Glass bottle
AC257740010 1 L Glass bottle
AC257740050 5 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC257740025 Supplier Thermo Scientific Chemicals Supplier No. 257740025
Only null left
Add to Cart

CAS: 3811-73-2 | C5H4NNaOS | 149.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Mercaptopyridine-N-oxide, sodium salt
Name Note 40 w/w% Aqueous Solution
CAS 3811-73-2, 7732-18-5
CAS Min % 40.0, 58.0
CAS Max % 42.0, 60.0
Melting Point -30.0°C
Color Amber to Yellow
Density 1.2200g/mL
pH 8.5 to 10.5 (10% soln. at 20°C)
Boiling Point 109.0°C
Packaging Glass bottle
Molecular Formula C5H4NNaOS
Quantity 2.5 L
MDL Number MFCD01941547
Specific Gravity 1.22
Merck Index 10, 7892
Synonym sodium omadine, sodium pyrithione, sodium 2-pyridylthio-n-oxide, pyrithione sodium salt, thione reagent, 2-pyridinethiol-1-oxide sodium salt, 2-mercaptopyridine n-oxide sodium salt, omadine sodium, sodium-2-pyridinethiol-1-oxide, sodium omadine van
Solubility Information Solubility in water: soluble
InChI Key XNRNJIIJLOFJEK-UHFFFAOYSA-N
SMILES C1=CC(=S)N(C=C1)[O-].[Na+]
IUPAC Name sodium;1-oxidopyridine-2-thione
Molecular Weight (g/mol) 149.15
PubChem CID 19658
Formula Weight 149.15
Percent Purity 40 to 42%
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.