Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl acetate, 99.5+%, for HPLC
GREENER_CHOICE

Catalog No. AC268350010
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC268350010 1 L
AC268350025 2.5 L
AC268350040 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC268350010 Supplier Thermo Scientific Chemicals Supplier No. 268350010
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 141-78-6, C4H8O2, 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Melting Point -83.5°C
Absorbance 1 cm cell vs HPLC-grade water, 1 cm cell vs HPLC-grade water at 0 nm, 0.005 max. at 280nm, 0.025 max. at 270nm, 0.12 max. at 260nm, 1 max. at 255nm
Color Colorless
Acidity 0.0015 meq/g max.
Density 0.9020g/mL
Quantity 1 L
Flash Point -4°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3710 to 1.3740
Residue after Evaporation 0.0002 % max.
Specific Gravity 0.902
Assay Percent Range 99.5+%
Linear Formula CH3CO2C2H5
Vapor Pressure Vapor Pressure: 103mbar at 20°C
Viscosity 0.45 mPa.s (20°C)
Water 0.03% Max.
Beilstein 02, 125
Merck Index 15, 3811
Green Features ACT Ecolabel (Silver)
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether
Boiling Point 75.0°C to 78.0°C
Formula Weight 88.11
Percent Purity ≥99.5%
Grade HPLC
Chemical Name or Material Ethyl acetate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-500-4
RTECSNumber AH5425000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.