Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydroxide, 1N solution in water

Catalog No. AC278400250
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Plastic Can
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC278400250 25 L Plastic Can
AC278400010 1 L Plastic bottle
AC278400025 2.5 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC278400250 Supplier Thermo Scientific Chemicals Supplier No. 278400250
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 1310-58-3 | HKO | 56.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium hydroxide
Color Colorless
Name Note 1N solution in water
Boiling Point 100.0°C
CAS 1310-58-3, 7732-18-5
Molecular Formula HKO
Linear Formula KOH
MDL Number MFCD00003553
Concentration 1N, exact strength on the certificate of analysis
UN Number 1814
Fieser 01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258
Density 1.0500g/mL
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Solubility in water: soluble.
Quantity 25 L
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11
Packaging Plastic Can
Specific Gravity 1.05
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.