Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals beta-D-Glucose pentaacetate, 98%

Catalog No. AC302961000
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC302961000 100 g Plastic bottle
AC302965000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AC302961000 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 302961000
Only null left
Add to Cart

CAS: 604-69-3 | C16H22O11 | 390.341 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-69-3
MDL Number MFCD00006597
InChI Key LPTITAGPBXDDGR-IBEHDNSVSA-N
Synonym beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-β
PubChem CID 2724702
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 129.0°C to 133.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Molecular Formula C16H22O11
Quantity 100 g
Specific Rotation Condition + 5.00 (20.00°C c=1,CHCL3)
Specific Rotation + 5.00
Molecular Weight (g/mol) 390.33
Formula Weight 390.33
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material β-D-Glucose pentaacetate
Show More Show Less
EINECSNumber 210-074-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.