Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc acetate dihydrate, 98+%, ACS reagent

Catalog No. AC317210025
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC317210025 2.5 kg Plastic bottle
AC317215000 500 g Plastic bottle
AC317210100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC317210025 Supplier Thermo Scientific Chemicals Supplier No. 317210025
Only null left
Add to Cart

CAS: 5970-45-6 | C4H6O4Zn·2H2O | 219.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H6O4Zn·2H2O
Molecular Weight (g/mol) 219.5
MDL Number MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Melting Point 237.0°C
Color White
pH 6.0 to 7.0 (5% soln. at 25°C)
Physical Form Crystals
Quantity 2.5 kg
Assay Percent Range 98.0 to 101% (Complexometry)
Linear Formula (CH3CO2)2Zn·2H2O
Merck Index 15,10327
Solubility Information Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 219.5
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 1.84
Density 1.8400g/mL
Chemical Name or Material Zinc acetate dihydrate, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously wit
RTECSNumber ZG8750000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.