Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-ornithine hydrochloride, 99%

Catalog No. AC347000250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC347000250 25 g
AC347000050 5 g
AC347001000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC347000250 Supplier Thermo Scientific Chemicals Supplier No. 347000250
Only null left
Add to Cart

CAS: 1069-31-4 | C5H13ClN2O2 | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1069-31-4
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.62
MDL Number MFCD00065398
InChI Key GGTYBZJRPHEQDG-UHFFFAOYNA-N
Synonym dl-ornithine hydrochloride, 2,5-diaminopentanoic acid hydrochloride, dl-ornithine monohydrochloride, dl-ornithinehcl, h-dl-orn-oh.hcl, dl-ornithine hcl, dl-ornithine, hydrochloride, ornithine monohydrochloride, ornithine hydrochloride, ornithine, hydrochloride 1:1
PubChem CID 71598
SMILES [H+].[Cl-].NCCCC(N)C(O)=O

Specifications

Melting Point 291°C
Color White
Loss on Drying 0.3% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2N(CH2)3CH(NH2)CO2H·HCl
Quantity 25 g
Solubility Information (10% in water) Clear colorless to light yellow solution
Transmittance 95% min. (c=5, H2O, 430nm) 1cm cell
IUPAC Name 2,5-diaminopentanoic acid;hydrochloride
Formula Weight 168.62
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-Ornithine hydrochloride, 99%
Show More Show Less
EINECSNumber 213-956-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.