Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TentaGel™ S-RAM

Catalog No. AC354650050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC354650050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC354650050 Supplier Thermo Scientific Chemicals Supplier No. 354650050
Only null left
Add to Cart
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For solid support synthesis
TRUSTED_SUSTAINABILITY

Specifications

Color White to Yellow
Chemical Name or Material TentaGel™ S-RAM
Physical Form Powder
Loading 0.2 to 0.3mmol/g
Packaging Glass bottle
Particle Size 90 Micron
Quantity 5 g
MDL Number MFCD00217896
Synonym N-FMOC-4'-[poly(oxyethylene)carbamoylmethoxy]-2, 4-dimethoxy-benzhydrylamine polymer bound, Poly(oxyethylene)-RAM polymer bound

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.