Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid, pure, ca. 32% solution in water

Catalog No. AC389310025
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC389310025 2.5 L Glass bottle
1 options

Catalog No. AC389310025

Supplier: Thermo Scientific Chemicals 389310025

Due to product restrictions, please to purchase or view availability for this product.

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless
Melting Point -35.0°C
Boiling Point 57.0°C
Density 1.1600g/mL
Heavy Metals (as Pb) 5ppm max.
Packaging Glass bottle
Residue after Ignition 0.005% max.
Specific Gravity 1.16
Quantity 2.5 L
Viscosity 1.9 mPa.s (15°C)
Physical Form Liquid
Linear Formula HCl
Merck Index 15, 4818
Formula Weight 36.45
Percent Purity ≥32%
Grade Pure
Chemical Name or Material Hydrochloric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be corrosive to metals. Causes severe skin burns and eye damage. May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.