Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, for HPLC

Catalog No. AC389390010
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC389390010 1 L Glass Bottle
AC389390025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC389390010 Supplier Thermo Scientific Chemicals Supplier No. 389390010
  • $122.60
    $104.65 / Each of 1

    Save $17.95

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7732-18-5 | H2O | 18.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance 0.004 max. at 254nm, 0.009 max. at 210nm, 0.05 max. at 200nm
Color Colorless
Boiling Point 100.0°C
Conductivity 1μS/cm Max.
Linear Formula H2O
Merck Index 15, 10237
Formula Weight 18.02
Grade HPLC
Packaging Glass Bottle
Residue after Evaporation 0.0003% max.
Viscosity 0.89 mPa.s (25°C)
Density 1.0000g/mL
Quantity 1 L
Refractive Index 1.3334
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Hazard Category
EINECSNumber 231-791-2
RTECSNumber ZC0110000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.