Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen chloride, ca. 0.5M solution in methanol

Catalog No. AC391741000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
Glass bottle

Catalog No. AC391741000

Supplier: Thermo Scientific Chemicals 391741000

Only null left
Add to Cart

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrochloric acid
Color Colorless
Name Note 0.5M solution in methanol
CAS 7647-01-0, 67-56-1
Concentration 0.4 to 0.6M
Density 1.0200g/mL
Packaging Glass bottle
Specific Gravity 1.02
Flash Point 12°C
Quantity 100 mL
Molecular Formula ClH
Physical Form Liquid
Linear Formula HCl
MDL Number MFCD00011324 MFCD00792839
Merck Index 14, 4794
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.