Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium phosphate, monobasic, 99+%, ACS reagent

Catalog No. AC424200250
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424200250 25 g Plastic bottle
AC424205000 500 g Plastic bottle
AC424200025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC424200250 Supplier Thermo Scientific Chemicals Supplier No. 424200250
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253.0°C
Color Colorless to White
pH 4.1 to 4.5 (5% soln. at 25°C)
Physical Form Crystals
Quantity 25 g
Loss on Drying 0.2% max. (2 g, 105°C, 2 hrs)
Packaging Plastic bottle
Decomposition Information >250°C
Assay Percent Range 99+%
Linear Formula KH2PO4
Merck Index 15, 7780
Solubility Information Solubility in water: 222g/L (20°C). Other solubilities: insoluble in alcohol
IUPAC Name potassium dihydrogen phosphate
Formula Weight 136.09
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Potassium phosphate, monobasic
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 231-913-4
RTECSNumber TC6615500
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.