Learn More
Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| pH | 8.7 to 9.3 (5% soln. at 25°C) |
| Physical Form | Granular Powder |
| Quantity | 10 kg |
| Assay Percent Range | 99+% |
| Linear Formula | Na2HPO4 |
| Merck Index | 15, 8789 |
| Solubility Information | Solubility in water: 75g/L (20°C). Other solubilities: practically insoluble in alcohol |
| IUPAC Name | disodium;hydrogen phosphate |
| Formula Weight | 141.96 |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: CAUTION! |
| Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Hygroscopic. NFPA Health:1 Flammability:0 Instability:1 |
| EINECSNumber | 231-448-7 |
| RTECSNumber | WC4500000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.