Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, 99.5%, ACS reagent, meets the requirements of Reag. Ph.Eur.

Catalog No. AC424555000
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424555000 500 mL EcoSafPak™ Glass Bottle
AC424550010 1 L EcoSafPak™ Glass Bottle
AC424550025 2.5 L Glass Bottle
AC424550251 25 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC424555000 Supplier Thermo Scientific Chemicals Supplier No. 424555000
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 108-88-3 | C7H8 | 92.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95.0°C
Density 0.8660g/mL
Quantity 500 mL
Flash Point 4°C
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.4968
Residue after Evaporation 0.001% max.
Specific Gravity 0.866
Assay Percent Range 99.5%
Vapor Pressure Vapor Pressure: 29mbar at 20°C
Linear Formula C6H5CH3
Viscosity 0.6 mPa.s (20°C)
Water 0.03% max. (Coulometric)
Beilstein 05, 280
Merck Index 15, 9688
Boiling Point 111.0°C
Solubility Information Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
Formula Weight 92.14
Percent Purity 99.5%
Grade ACS
Chemical Name or Material Toluene
Show More Show Less
Hazard Category Signal Word: DANGER!
Hazard Statement GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.