Learn More
Potassium tert-butoxide, 1M solution in tert-butanol, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 800 mL |
| Linear Formula | (CH3)3COK |
| Flash Point | 12°C |
| Formula Weight | 112.21 |
| Concentration or Composition (by Analyte or Components) | 0.90 to 1.10M |
| Packaging | AcroSeal™ Glass bottle |
| Appearance | Colorless to amber solution |
| Specific Gravity | 0.82 |
| Density | 0.8200g/mL |
| Chemical Name or Material | Potassium tert-butoxide, 1M solution in tert-butanol |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Danger |
| missing translation for 'healthHazard2' | GHS H Statement Highly flammable liquid and vapor. Harmful if inhaled. Causes severe skin burns and eye damage. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact |
| EINECSNumber | 212-740-3 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.