Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane, 95+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number AC445780025
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
CCCCCC
This item is not returnable. View return policy
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
CCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -95.0°C
Density 0.6590 g/mL
Boiling Point 69.0°C
Quantity 2.5 L
Flash Point -22°C
Refractive Index 1.3748 to 1.3810
Assay Percent Range 95+%
Specific Gravity 0.659
Linear Formula CH3(CH2)4CH3
Viscosity 0.31 mPa.s (20°C)
Merck Index 15, 4732
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Formula Weight 86.18
Grade ACS Reagent
Chemical Name or Material n-Hexane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May be fatal if swallowed and enters airways.

May cause drowsiness or dizziness.

Causes skin irritation.

May cause damage to organs through prolonged or repeated exposure.

Toxic to aquatic life with long las
Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

Use personal protective equipment as required.

Do not breathe dust/fume/gas/mist/vapors/spray.

IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

Do NOT induce vomiting.

Avoid release to the environment.

IF ON SKIN: Wash with plenty of soap and water.

EINECSNumber 203-777-6

RUO – Research Use Only

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit