Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Isopropanol, 99%, for biochemistry and histology, AcroSeal™ Catalog No.: AC447080010
Only null left
Add to Cart

Isopropanol, 99%, for biochemistry and histology, AcroSeal™

Catalog No. AC447080010
1 L, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle

Catalog No. AC447080010

Supplier: Thermo Scientific Chemicals 447080010

Only null left
Add to Cart
Only null left
Add to Cart
Request Bulk or Custom Formats

CAS: 67-63-0 | C3H8O | 60.096 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petroholShow MoreShow Less
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -89.5°C
Absorbance 0.01 max. at 300nm, 0.03 max. at 260nm, 0.05 max. at 240nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 1 max. at 205nm
Density 0.7850g/mL
Boiling Point 81.0°C to 83.0°C
Quantity 1 L
Flash Point 12°C
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3760 to 1.3780
Assay Percent Range 99%
Specific Gravity 0.785
Linear Formula (CH3)2CHOH
Viscosity 2.27 mPa.s (20°C)
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride,, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
Formula Weight 60.1
Percent Purity 99%
Grade Biochemical
Chemical Name or Material Isopropanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2538064 Certificate TypeCertificate of Analysis Date05/24/2025 Catalog Number447080010,447080025
Lot Number2531116 Certificate TypeCertificate of Analysis Date05/24/2025 Catalog Number447080010,447080025
Lot Number2532234 Certificate TypeCertificate of Analysis Date05/24/2025 Catalog Number447080010,447080025
Lot Number2530510 Certificate TypeCertificate of Analysis Date05/24/2025 Catalog Number447080010,447080025
Lot Number2528863 Certificate TypeCertificate of Analysis Date05/24/2025 Catalog Number447080010,447080025
Hazard Category Danger

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isopropanol, 99%, for biochemistry and histology, AcroSeal™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.