Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen chloride, technical, 4 to 6N solution in 2-propanol

Catalog No. AC450720010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450720010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC450720010 Supplier Thermo Scientific Chemicals Supplier No. 450720010
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrochloric acid
CAS 7647-01-0, 67-63-0
Molecular Formula ClH
Linear Formula HCl
MDL Number MFCD00011324 MFCD00792839
Density 0.9090g/mL
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Solubility in water: >1000g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol: >50g/L
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.45
Grade Technical
Packaging Glass bottle
Specific Gravity 0.909
Quantity 1 L
Flash Point 11°C
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.